Learn More
Vici Metronics Sulfur Traps, 1/8'' Fitting Size, 12'' Length
SDP
Catalog No. NC1568104
Encompass_Preferred
Removes all sulfur-containing compounds from any non-reactive gas. Also removes halocarbons and most non-methane hydrocarbons.
Catalog No. NC1568104 Supplier Vici Metronics Supplier No. T4001
Removes all sulfur-containing compounds from any non-reactive gas. Also removes halocarbons and most non-methane hydrocarbons.