Learn More
Ace Glass, Inc. Trap, bump, 250mL, 14/20 top, 14/20 bottom
SDP
Catalog No. 50973367
Encompass_Preferred
250mL bump trap for use between the rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 24/40 top and bottom joints.
Catalog No. 50-973-367 Supplier Ace Glass, Inc. Supplier No. 670414
250mL bump trap for use between the rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 24/40 top and bottom joints.