Learn More
Ace Glass, Inc. Trap, bump, drain holes, 250mL, 14/20 top, 14/20 bottom
SDP
Catalog No. 50973368
Encompass_Preferred
250mL bump trap for use between a rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 14/20 top and bottom joints. Improved design incorporates two drain holes to expedite the return of solvent to the evaporator flask.
Catalog No. 50-973-368 Supplier Ace Glass, Inc. Supplier No. 670512
250mL bump trap for use between a rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 14/20 top and bottom joints. Improved design incorporates two drain holes to expedite the return of solvent to the evaporator flask.