Learn More
Ace Glass, Inc. Trap, bump, 250mL, 29/42 top, 29/42 bottom
SDP
Catalog No. 50978862
Encompass_Preferred
250mL bump trap for use between the rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 29/42 top and bottom joints.
Catalog No. 50-978-862 Supplier Ace Glass, Inc. Supplier No. 670420
250mL bump trap for use between the rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 29/42 top and bottom joints.