Learn More
Ace Glass, Inc. Trap, bump, drain holes, 250mL, 29/42 top, 29/42 bottom
SDP
Catalog No. 50973369
Encompass_Preferred
250mL bump trap for use between a rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 29/42 top and bottom joints. Improved design incorporates two drain holes to expedite the return of solvent to the evaporator flask.
Catalog No. 50-973-369 Supplier Ace Glass, Inc. Supplier No. 670522
250mL bump trap for use between a rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 29/42 top and bottom joints. Improved design incorporates two drain holes to expedite the return of solvent to the evaporator flask.