Learn More
Cole-Parmer Chromatography Research Supplies Carbon Dioxide Traps; 40 liters volume; 1/4" fittings
Catalog No. NC1797951 Shop All Cole-Parmer Products
Encompass
Traps remove carbon dioxide from your gas stream. A 500-mL nickel-plated carbon steel cylinder holds packed absorbent (80% emulsive, 320 g; 20% NaOH, 70 g); withstands up to 1000 psi. Include drying stage to prevent moisture from entering system. If downstream moisture is harmful to application, a second drying stage is recommended. Trap measures 15"L x 2" dia; frits are integral to each fitting. Regeneration or refilling is not recommended. Shpg wt 3 lb (1.4 kg).
Catalog No. NC1797951 Supplier Cole-Parmer Supplier No. 3465170
Traps remove carbon dioxide from your gas stream. A 500-mL nickel-plated carbon steel cylinder holds packed absorbent (80% emulsive, 320 g; 20% NaOH, 70 g); withstands up to 1000 psi. Include drying stage to prevent moisture from entering system. If downstream moisture is harmful to application, a second drying stage is recommended. Trap measures 15"L x 2" dia; frits are integral to each fitting. Regeneration or refilling is not recommended. Shpg wt 3 lb (1.4 kg).