Learn More
Ace Glass, Inc. Trap cold 40mm ID 250mm lg 35/25-40mm ID O-ring joints
SDP
Catalog No. 50979479
Encompass_Preferred
250mm Liquid Nitrogen Trap with 35/25 o-ring joints, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-979-479 Supplier Ace Glass, Inc. Supplier No. 781850
250mm Liquid Nitrogen Trap with 35/25 o-ring joints, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.