Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Trap Column, nanoEase M/Z, 5 μm, 100 Å

Catalog No. NC3958476 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These nanoEase M/Z HSS T3 Trap Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation. They offer high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. The columns withstand 15,000 psi operating pressure, ensuring superior longevity.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45 °C
  • Maximum pressure: 15,000 psi (1034 Bar)
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 100 Å
  • Inner diameter: 300 μm
  • Length: 50 mm
  • Bonding technology: T3
  • Particle technology: HSS
  • QC tested: Peptide

Catalog No. NC3958476 Supplier Waters Corp Supplier No. 186009266
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These nanoEase M/Z HSS T3 Trap Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation. They offer high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. The columns withstand 15,000 psi operating pressure, ensuring superior longevity.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45 °C
  • Maximum pressure: 15,000 psi (1034 Bar)
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 100 Å
  • Inner diameter: 300 μm
  • Length: 50 mm
  • Bonding technology: T3
  • Particle technology: HSS
  • QC tested: Peptide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.