Learn More
Perkin Elmer US LLC In-Line Indicating Oxygen Trap
Catalog No. 509045784
Encompass_Preferred
This high-efficiency indicator trap reduces oxygen levels to less than 0.1 ppm. Changes color from bright green to gray when adsorption capacity is depleted. Oxygen capacity for this compact unit is 0.05 g at STP. The non-contaminating, heavy-wall inner glass tube of adsorbent is protected from breakage by the outer plastic tube Fitting Size: 1/4 inch (Brass) Maximum Pressure: 200 psi Dimensions: 2 in x 20 in (including fittings) Weight 3.5 lb/1.6 kg Recommended Flow Rate: Up to 2 L/min
Catalog No. 50-904-5784 Supplier Perkin Elmer US LLC Supplier No. N9301191
This high-efficiency indicator trap reduces oxygen levels to less than 0.1 ppm. Changes color from bright green to gray when adsorption capacity is depleted. Oxygen capacity for this compact unit is 0.05 g at STP. The non-contaminating, heavy-wall inner glass tube of adsorbent is protected from breakage by the outer plastic tube Fitting Size: 1/4 inch (Brass) Maximum Pressure: 200 psi Dimensions: 2 in x 20 in (including fittings) Weight 3.5 lb/1.6 kg Recommended Flow Rate: Up to 2 L/min