Learn More
Wilmad Labglass TRAP MOISTURE B-S 5ML 24/40
Supplier:  Wilmad Labglass LG9100100
Encompass
Moisture Trap, Bidwell-Sterling, w/ Overflow Tube. Capacity: 5mL; Subdivision: 0.10mL; Joint Size: [ST] 24/40. Design allows excess solvent to return to the boiling flask which minimizes the chance of return of condensate.
Catalog No. 50-806-797