Learn More
Wilmad Labglass TRAP MOISTURE B-S 5ML 24/40
Supplier:  Wilmad Labglass LG9095100
Encompass
Moisture Trap, Bidwell-Sterling. Capacity: 5mL; Subdivision: 0.10mL; Joint Size: [ST] 24/40. Calibrated to contain at 20degreeC. For determination of water in materials by distillation w/ a solvent that is not miscible w/ water.
Catalog No. 50-804-478