Learn More
Wilmad Labglass TRAP MOISTURE BARRT 20ML 24/40
Catalog No. 50803734
Encompass
Moisture Trap, Barrett, w/ Glass Stopcock. Capacity: 20mL; Graduations: 0-3mL: 0.2mL, 3-20mL: 0.5mL; Joint Size: [ST] 24/40. Calibrated to contain at 20degreeC.
Catalog No. 50-803-734 Supplier Wilmad Labglass Supplier No. LG9075102
Add to Cart
Add to Cart
Moisture Trap, Barrett, w/ Glass Stopcock. Capacity: 20mL; Graduations: 0-3mL: 0.2mL, 3-20mL: 0.5mL; Joint Size: [ST] 24/40. Calibrated to contain at 20degreeC.