Learn More
Perkin Elmer US LLC Trap Packing Disc, Pkg. 20
Catalog No. 502112798
Encompass_Preferred
Glass fiber disk tor separating sorbents when packing ATD cold trap with multiple sorbents. Recommended when using low flow trap.
Catalog No. 50-211-2798 Supplier Perkin Elmer US LLC Supplier No. L4271290
Glass fiber disk tor separating sorbents when packing ATD cold trap with multiple sorbents. Recommended when using low flow trap.