Learn More
Chemglass Life Sciences Trap to fit AF-0350-01 90L/min Pump, Airfree, Schlenk
SDP
Catalog No. 501530948
Encompass_Preferred
Trap to fit AF-0350-01 90L/min Pump, Airfree, Schlenk. Designed to fit between the vacuum pump and the upper cart shelf on the AF-0300-01 cart to prevent oil from being drawn into the vacuum line. Offset inlet and outlet permits installation as illustrated. All sizes of the trap are fitted with a 4mm Airfree valve to allow the equalization of pressure when the pump is shut down while still having the line portion remain evacuated. Arms on all sizes have 1" OD glass tubing. Requires two AF-0300-11 hose clamps for installation.
Catalog No. 50-153-0948 Supplier Chemglass Life Sciences Supplier No. AF030008
Trap to fit AF-0350-01 90L/min Pump, Airfree, Schlenk. Designed to fit between the vacuum pump and the upper cart shelf on the AF-0300-01 cart to prevent oil from being drawn into the vacuum line. Offset inlet and outlet permits installation as illustrated. All sizes of the trap are fitted with a 4mm Airfree valve to allow the equalization of pressure when the pump is shut down while still having the line portion remain evacuated. Arms on all sizes have 1" OD glass tubing. Requires two AF-0300-11 hose clamps for installation.