Learn More
Wilmad Labglass TRAP ROT-EVP 100ML 24/40 24/40
Catalog No. 50807347
Encompass_Preferred
Trap, Rotary Evaporator. Size: 100mL; Upper Joint Size: [ST] 24/40 Outer; Lower Joint Size: [ST] 24/40 Inner. Reduces foam carry-over in Rotary Evaporator s. Trap is used between vapor duct tube and evaporator flask.
Catalog No. 50-807-347 Supplier Wilmad Labglass Supplier No. LG6880104
Trap, Rotary Evaporator. Size: 100mL; Upper Joint Size: [ST] 24/40 Outer; Lower Joint Size: [ST] 24/40 Inner. Reduces foam carry-over in Rotary Evaporator s. Trap is used between vapor duct tube and evaporator flask.