Learn More
Wilmad Labglass TRAP ROT-EVP 250ML 24/40 14/20
Supplier:  Wilmad Labglass LG6880106
Encompass_Preferred
Trap, Rotary Evaporator. Size: 250mL; Upper Joint Size: [ST] 24/40 Outer; Lower Joint Size: [ST] 14/20 Inner. Reduces foam carry-over in Rotary Evaporator s. Trap is used between vapor duct tube and evaporator flask.
Catalog No. 50-807-348