Learn More
Wilmad Labglass TRAP ROT-EVP 250ML 24/40 24/40
Supplier:  Wilmad Labglass LG6883104
Encompass_Preferred
Trap. Rotary Evaporator, Improved. Size: 250mL; Upper Joint Size: [ST] 24/40 Outer; Lower Joint Size: [ST] 24/40 Inner. Improved pear-shaped design facilitates the return of condensate to the evaporator flask.
Catalog No. 50-805-655