Learn More
Wilmad Labglass TRAP ROT-EVP 250ML 24/40 24/40
Catalog No. 50806660
Encompass_Preferred
Trap, Rotary Evaporator. Size: 250mL; Upper Joint Size: [ST] 24/40 Outer; Lower Joint Size: [ST] 24/40 Inner. Reduces foam carry-over in Rotary Evaporator s. Trap is used between vapor duct tube and evaporator flask.
Catalog No. 50-806-660 Supplier Wilmad Labglass Supplier No. LG6880110
Trap, Rotary Evaporator. Size: 250mL; Upper Joint Size: [ST] 24/40 Outer; Lower Joint Size: [ST] 24/40 Inner. Reduces foam carry-over in Rotary Evaporator s. Trap is used between vapor duct tube and evaporator flask.