Learn More
Wilmad Labglass TRAP ROT-EVP 250ML 24/40 24/40
Supplier:  Wilmad Labglass LG6881110
Encompass_Preferred
Trap, Rotary Evaporator, Drain Back. Size: 250mL; Upper Joint Size: [ST] 24/40 Outer; Lower Joint Size: [ST] 24/40 Inner. Base of inner tube has two 3mm openings to allow drain-back of foam condensate to the evaporator flask.
Catalog No. 50-805-829