Learn More
Cosmo Bio Usa Inc TRAP Staining Kit, 10 kits, 50 ml Tartrate-containing Buffer, 10 vials Chromogenic Substrate, for staining osteoclasts, 96-well plate format
SDP
Supplier:  Cosmo Bio Usa Inc PMCAK04F
Encompass
TRAP Staining Kit, 10 kits, 50 ml Tartrate-containing Buffer, 10 vials Chromogenic Substrate, for staining osteoclasts, 96-well plate format
Catalog No. NC2256842