Learn More
Wilmad Labglass TRAP VAC 2-PC 28X200MM COMP
Supplier:  Wilmad Labglass LG11020098
Encompass_Preferred
Vacuum Trap, Two-Piece, Straight Top Stem, Complete. Body O.D.: 28mm; Body Length: 200mm; Joint: [ST] 24/40; Stem O.D.: 10mm. Body and Stopper connected by standard taper joint. Both Inlet and outlet stems are integral with the Stopper.
Catalog No. 50-807-522