Learn More
Wilmad Labglass TRAP VAC 2-PC 32X200MM COMP
Supplier:  Wilmad Labglass LG11020100
Encompass_Preferred
Vacuum Trap, Two-Piece, Straight Top Stem, Complete. Body O.D.: 32mm; Body Length: 200mm; Joint: [ST] 29/42; Stem O.D.: 10mm. Body and Stopper connected by standard taper joint. Both Inlet and outlet stems are integral with the Stopper.
Catalog No. 50-805-623