Learn More
Wilmad Labglass TRAP VAC 2-PC 38X225MM COMP
Supplier:  Wilmad Labglass LG11020102
Encompass_Preferred
Vacuum Trap, Two-Piece, Straight Top Stem, Complete. Body O.D.: 38mm; Body Length: 225mm; Joint: [ST] 34/45; Stem O.D.: 13mm. Body and Stopper connected by standard taper joint. Both Inlet and outlet stems are integral with the Stopper.
Catalog No. 50-803-672