Learn More
Wilmad Labglass TRAP VAC 2-PC 48X250MM COMP
Supplier:  Wilmad Labglass LG11020106
Encompass_Preferred
Vacuum Trap, Two-Piece, Straight Top Stem, Complete. Body O.D.: 48mm; Body Length: 250mm; Joint: [ST] 45/50; Stem O.D.: 20mm. Body and Stopper connected by standard taper joint. Both Inlet and outlet stems are integral with the Stopper.
Catalog No. 50-804-193
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.