Learn More
Wilmad Labglass TRAP VAC 2PC 32X200MM COMP
Catalog No. 50806589
Encompass_Preferred
Vacuum Trap, Two-Piece, with Hose Connectors, Complete. Body O.D.: 32mm; Body Length: 200mm; Joint: [ST] 29/42; Hose Connectors: 3/8 in.. For removal of vapors from a vacuum line by condensation with liquid air or dry ice slurry mixture
Catalog No. 50-806-589 Supplier Wilmad Labglass Supplier No. LG11026100
Vacuum Trap, Two-Piece, with Hose Connectors, Complete. Body O.D.: 32mm; Body Length: 200mm; Joint: [ST] 29/42; Hose Connectors: 3/8 in.. For removal of vapors from a vacuum line by condensation with liquid air or dry ice slurry mixture