Learn More
Ace Glass, Inc. Vacuum trap, No-Air™;, 40mm (65/40) spherical o-ring flange, 15mm O-ring joint arms. 19mm OD vacuum stem, 45mm OD body with 210mm overall length. 7669-20 stainless pinch clamp and size -226 Viton; O-ring included.
SDP
Catalog No. 50976402
Encompass_Preferred
Vacuum trap, No-Air™;, 40mm (65/40) spherical o-ring flange, 15mm O-ring joint arms. 19mm OD vacuum stem, 45mm OD body with 210mm overall length. 7669-20 stainless pinch clamp and size -226 Viton; O-ring included.
Catalog No. 50-976-402 Supplier Ace Glass, Inc. Supplier No. 8771100
Vacuum trap, No-Air™;, 40mm (65/40) spherical o-ring flange, 15mm O-ring joint arms. 19mm OD vacuum stem, 45mm OD body with 210mm overall length. 7669-20 stainless pinch clamp and size -226 Viton; O-ring included.