Learn More
Wilmad Labglass TRAP VACUUM 1-PC 60X250MM 1/4
Catalog No. 50804313
Encompass_Preferred
Vacuum Trap, One Piece. Body O.D.: 60mm; Body Length: 250mm; Hose Connections: 1/4 in.. One piece design having two vertical Hose Connections. Design affords use with a wide variety of Dewar flasks.
Catalog No. 50-804-313 Supplier Wilmad Labglass Supplier No. LG11017100
Vacuum Trap, One Piece. Body O.D.: 60mm; Body Length: 250mm; Hose Connections: 1/4 in.. One piece design having two vertical Hose Connections. Design affords use with a wide variety of Dewar flasks.