Learn More
Ace Glass, Inc. 50/50 Vacuum Trap w/ Bent Vertical Hose Barb Stopper
SDP
Catalog No. 50976384
Encompass_Preferred
Bent vertical hose barb vacuum trap with stopper with 50/50 joint. Hose barb for 7/16 or 1/2 tubing. Outer body dimensions: 54mm O.D. x 110mm length.
Catalog No. 50-976-384 Supplier Ace Glass, Inc. Supplier No. 876074
Bent vertical hose barb vacuum trap with stopper with 50/50 joint. Hose barb for 7/16 or 1/2 tubing. Outer body dimensions: 54mm O.D. x 110mm length.