Learn More
Ace Glass, Inc. 55/60 Vacuum Trap w/ Bent Vertical Hose Barb Stopper
SDP
Catalog No. 50979867
Encompass_Preferred
Bent vertical hose barb vacuum trap with stopper with 55/50 joint. Hose barb for 7/16 or 1/2 tubing. Outer body dimensions: 60mm O.D. x 140mm length.
Catalog No. 50-979-867 Supplier Ace Glass, Inc. Supplier No. 876064
Bent vertical hose barb vacuum trap with stopper with 55/50 joint. Hose barb for 7/16 or 1/2 tubing. Outer body dimensions: 60mm O.D. x 140mm length.