Learn More
Wilmad Labglass TRAP VACUUM DEWAR 250ML
Supplier:  Wilmad Labglass LG11065094
Encompass
Vacuum Trap, Dewar Shaped. Inner Chamber I.D.: 59mm; Inner Chamber Depth: 230mm; Reservoir Capacity: 250mL; PTFE Valve Size: 0-4mm. Designed to remove relatively large amounts of solvent
Catalog No. 50-805-296