Learn More
Ace Glass, Inc. Trap, vacuum, complete, bent vertical tube, 34/45, 38mm outer body OD, 250mm outer body length below joint, 16mm inner & side tube OD
SDP
Catalog No. 50976359
Encompass_Preferred
Trap, vacuum, complete, bent vertical tube, 34/45, 38mm outer body OD, 250mm outer body length below joint, 16mm inner & side tube OD
Catalog No. 50-976-359 Supplier Ace Glass, Inc. Supplier No. 875343
Trap, vacuum, complete, bent vertical tube, 34/45, 38mm outer body OD, 250mm outer body length below joint, 16mm inner & side tube OD