Learn More
Ace Glass, Inc. Trap, vacuum, complete, bent vertical tube, 40/50, 45mm outer body OD, 250mm outer body length below joint, 19mm inner & side tube OD
SDP
Catalog No. 50976360
Encompass_Preferred
Trap, vacuum, complete, bent vertical tube, 40/50, 45mm outer body OD, 250mm outer body length below joint, 19mm inner & side tube OD
Catalog No. 50-976-360 Supplier Ace Glass, Inc. Supplier No. 875349
Trap, vacuum, complete, bent vertical tube, 40/50, 45mm outer body OD, 250mm outer body length below joint, 19mm inner & side tube OD