Learn More
Aviva Systems Biology Corp TRAP1 Antibody - N-terminal region (AVARP00012_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP00012_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAP1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TRAP1. Alternate names: HSP75, HSP90LApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-62448