Learn More
Enzo Life Sciences TRAP1 (human), (recombinant) (His-tag) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP848200
Encompass_Preferred
Alternative name: HSP75, Mitochondrial HSP90, MtHSP90. Endotoxin: <50EU/mg purified protein (LAL test). Purity: ≥95% (SDS-PAGE; Western blot). MW: ~72kDa. Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 150mM sodium chloride, 1mM DTT, and 1mM EDTA. Source: Produced in E. coli. Human TRAP1 (aa 60-704) is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7649