Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TRAP1 Human GST 50ug
SDP

Catalog No. 5000196619
Encompass_Preferred

Recombinant human TRAP1 protein comprising amino acids S60-E308 is supplied as an N-terminal GST-tagged fragment expressed in Escherichia coli. The lyophilized preparation is formulated from PBS with 6% trehalose (pH 7.4), has an apparent molecular weight of ~54.5 kDa, and is ≥90% pure by reducing SDS-PAGE. Store at -20°C; after reconstitution, store at 4°C for up to one week or -20°C for extended storage and avoid repeated freeze-thaw cycles.

  • Recombinant fragment spans S60-E308, representing the TRAP1 functional region.
  • N-terminal GST tag enables affinity purification and detection.
  • Expressed in Escherichia coli for bacterial expression yield.
  • Lyophilized formulation with trehalose enhances long-term stability.
  • High purity (≥90%) suitable for biochemical assays.
  • Recommended storage conditions minimize degradation after reconstitution.

Catalog No. 50-001-96619 Supplier Medchemexpress LLC Supplier No. HYP7153750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human TRAP1 protein comprising amino acids S60-E308 is supplied as an N-terminal GST-tagged fragment expressed in Escherichia coli. The lyophilized preparation is formulated from PBS with 6% trehalose (pH 7.4), has an apparent molecular weight of ~54.5 kDa, and is ≥90% pure by reducing SDS-PAGE. Store at -20°C; after reconstitution, store at 4°C for up to one week or -20°C for extended storage and avoid repeated freeze-thaw cycles.

  • Recombinant fragment spans S60-E308, representing the TRAP1 functional region.
  • N-terminal GST tag enables affinity purification and detection.
  • Expressed in Escherichia coli for bacterial expression yield.
  • Lyophilized formulation with trehalose enhances long-term stability.
  • High purity (≥90%) suitable for biochemical assays.
  • Recommended storage conditions minimize degradation after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.