Learn More
Enzo Life Sciences TRAP1 monoclonal antibody (9B6) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA9710050
Encompass_Preferred
Clone: 9B6. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human TRAP1. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7588