Learn More
Cayman Chemical TRAP6 amIDtrIfluoROCTAt 100mg
SDP
Catalog No. 502914810
Encompass_Preferred
A peptide PAR1 agonist; induces platelet aggregation in vitro (EC50 = 0.15 µM); induces relaxation of isolated guinea pig internal anal sphincter strips at 30 µM; increases cigarette smoke extract-induced IL-8 release from isolated human bronchial epithelial cells at 50 µM
Catalog No. 50-291-4810 Supplier Cayman Chemical Supplier No. 36881100 MG
A peptide PAR1 agonist; induces platelet aggregation in vitro (EC50 = 0.15 µM); induces relaxation of isolated guinea pig internal anal sphincter strips at 30 µM; increases cigarette smoke extract-induced IL-8 release from isolated human bronchial epithelial cells at 50 µM