Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Trapezoid Science Table
SCI_ED

Catalog No. S42391 Shop All Diversified Spaces Products
Change view
Click to view available options
Top Style:
ChemGuard
Plastic laminate
Dimensions (L x W x H):
24 x 60 x 36 in.
30 x 60 x 36 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Top Style Dimensions (L x W x H)
S42391 Plastic laminate 30 x 60 x 36 in.
S42393 ChemGuard 30 x 60 x 36 in.
S42395 Plastic laminate 24 x 60 x 36 in.
S42397 ChemGuard 24 x 60 x 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S42391 Supplier Diversified Spaces™ Supplier No. TZ714LBBK36C
Add to Cart
Add to Cart

With steel frame supporting the apron.

  • Sturdy construction with 2.25 in. solid legs
  • Steel frame supporting apron
  • Chemical-resistant, non-emitting UV finish

Specifications

Product Type Trapezoid Science Table
With Casters Yes
Color Oak
Autoclavable Not Autoclavable
No. of Shelves None
No. of Doors None
No. of Drawers None
Top Style Plastic laminate
Footrest No
With Tilt No
Includes Solid oak apron
Frame Material Steel
Dimensions (L x W x H) 30 x 60 x 36 in.
Finish Chemical-resistant, non-emitting UV
Material Oak
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.