Learn More
Aviva Systems Biology Corp TRAPPC2L Antibody - N-terminal region (ARP56852_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56852_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAPPC2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TRAPPC2L. Alternate names: HSPC176, MGC111156ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62012