Learn More
Aviva Systems Biology Corp TRAPPC5 Antibody - N-terminal region (ARP53292_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAPPC5. It was validated on Western Blot. TRAPPC5,trafficking protein particle complex 5,TRS31; MGC52424; TRAPPC5,trafficking protein particle complex subunit 5,trafficking protein particle complex subunit 5The immunogen is a synthetic peptide directed towards the N terminal region of human TRAPPC5. Alternate names: MGC52424, TRS31ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-822