Learn More
Aviva Systems Biology Corp TRAPPC5 Antibody - middle region (ARP53293_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53293_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRAPPC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRAPPC5. Alternate names: MGC52424, TRS31ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-60991