Learn More
Cyclodextrin Technologies Development TRAPPSOL HYDRO BETACYC 200GM
Catalog No. NC1166115
Encompass
Beta Cyclodextrin derivative for enhanced solubility >50g/100ml - meets USP/EP standards
Catalog No. NC1166115 Supplier Cyclodextrin Technologies Development Supplier No. THPBP200G
Beta Cyclodextrin derivative for enhanced solubility >50g/100ml - meets USP/EP standards