Learn More
Cyclodextrin Technologies Development Hydroxypropyl BCD, 100g
Catalog No. NC0999732
Encompass
Beta Cyclodextrin derivative for enhanced solubility >50g/100ml - meets USP/EP standards. Pharmaceutical Grade
Catalog No. NC0999732 Supplier Cyclodextrin Technologies Development Supplier No. THPBP100G
Beta Cyclodextrin derivative for enhanced solubility >50g/100ml - meets USP/EP standards. Pharmaceutical Grade