Learn More
Cyclodextrin Technologies Development Hydroxypropyl BCD, 300g
Catalog No. NC1308912
Encompass
Beta Cyclodextrin derivative for enhanced solubility >50g/100ml - meets USP/EP standards. Pharmaceutical Grade
Catalog No. NC1308912 Supplier Cyclodextrin Technologies Development Supplier No. THPBP300G
Beta Cyclodextrin derivative for enhanced solubility >50g/100ml - meets USP/EP standards. Pharmaceutical Grade