Learn More
Grainger Mint-X Rodent-Repellent Trash Bags, 60 gal, Black, 150/Pk.
Catalog No. 502751863
Encompass_Preferred
Mint-X Rodent-Repellent Trash Bags provide an effective, EPA-registered solution for deterring rats, mice, raccoons, and squirrels. These high-density polyethylene bags feature patented mint technology with essential oils, including Methyl Salicylate, embedded directly into the plastic to provide a long-lasting, consistent mint scent.
  • Repels rats, mice, raccoons, and squirrels
  • EPA-registered Category IV pesticide
  • Includes essential oils like Methyl Salicylate and Camphor Oil
  • Features 60 gal capacity with 50 lb max. load strength
  • Provides long-lasting scent that does not fade after one use
  • Designed for food service applications
Catalog No. 50-275-1863 Supplier Grainger Supplier No. 14L026
Mint-X Rodent-Repellent Trash Bags provide an effective, EPA-registered solution for deterring rats, mice, raccoons, and squirrels. These high-density polyethylene bags feature patented mint technology with essential oils, including Methyl Salicylate, embedded directly into the plastic to provide a long-lasting, consistent mint scent.
  • Repels rats, mice, raccoons, and squirrels
  • EPA-registered Category IV pesticide
  • Includes essential oils like Methyl Salicylate and Camphor Oil
  • Features 60 gal capacity with 50 lb max. load strength
  • Provides long-lasting scent that does not fade after one use
  • Designed for food service applications