Learn More
ACL Inc TRASH CAN DISSIPATIVE 5/CS
Catalog No. 501037919
Encompass_Preferred
High impact, non black container made of high-density polyethylene. Contains no carbon, dyes or additives, making it ideal for all applications demanding cleanliness. Ideal for Clean Rooms, manufacturing, inspection, quality control, and research industries. Measures 13 in. diameter x 21.75 in. high (11 gal )
Catalog No. 50-103-7919 Supplier ACL Inc Supplier No. 5075
High impact, non black container made of high-density polyethylene. Contains no carbon, dyes or additives, making it ideal for all applications demanding cleanliness. Ideal for Clean Rooms, manufacturing, inspection, quality control, and research industries. Measures 13 in. diameter x 21.75 in. high (11 gal )