Learn More
ACROBiosystems ACROBiosystems Anti-Trastuzumab Antibodies (recommended for PK/PD)
Supplier:  ACROBiosystems TRBY5B1MG
Encompass_Preferred
Antibody;1MG;IgG1/kappa;Recognizes Trastuzumab specifically, no cross reactivity with other humanized antibodies.;Anti-Trastuzumab Antibodies (recommended for PK/PD) (TRB-Y5b) are expressed from human 293 cells (HEK293).; ;Immobilized Trastuzumab at 1 μg/mL (100 μL/well) can bind Anti-Trastuzumab Antibodies (recommended for PK/PD) (Cat. No. TRB-Y5b) with a linear range of 0.19-6.25 ng/mL.
Catalog No. 50-195-6589