Learn More
Medchemexpress LLC HY-138298A 5mg , Trastuzumab deruxtecan CAS:1826843-81-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY138298A5MG
Encompass_Preferred
HY-138298A 5mg Trastuzumab deruxtecan CAS: 1826843-81-5 Trastuzumab deruxtecan (DS-8201a) is an anti-human epidermal growth factor receptor 2 (HER2) antibody-drug conjugate (ADC). Trastuzumab deruxtecan is composed of a humanized anti-HER2 antibody, an enzymatically cleavable peptide-linker, and a topoisomerase I inhibitor. Trastuzumab deruxtecan can be used for the research of HER2-positive breast cancer and gastric cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0108