Learn More
IBL America Trastuzumab (Herceptin) ELISA
Supplier:  IBL America TM09013
Encompass_Preferred
Enzyme immunoassay (ELISA) test kit for the determination of free Trastuzumab (Herceptin, Herclon) in serum and plasma. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0397