Learn More
Medchemexpress LLC HY-P9921 1mg , Trastuzumab emtansine CAS:1018448-65-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99211MG
Encompass_Preferred
HY-P9921 1mg Trastuzumab emtansine CAS: 1018448-65-1 Trastuzumab emtansine (Ado-Trastuzumab emtansine) is an antibody-drug conjugate (ADC) that incorporates the HER2-targeted antitumor properties of trastuzumab with the cytotoxic activity of the microtubule-inhibitory agent DM1 (derivative of maytansine). Trastuzumab emtansine can be used for the research of advanced breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9070